Order Entry
Italy
ContactUsLinkComponent
 
Irritant
4-(4-Chlorophenoxy)benzaldehyde ≥98.0% (by GC)
Catalog # TCIAC2006-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4-(4-Chlorophenoxy)benzaldehyde ≥98.0% (by GC)
Catalog # TCIAC2006-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    53.0 to 57.0 deg-C

Specification Test Results

About this item

 :
 :232.67 g/mol
Boiling Pt:181°C
 :GHS07
CAS Number:61343-99-5
Formula:C₁₃H₉ClO₂
MDL Number:MFCD01631895