Order Entry
Italy
ContactUsLinkComponent
4'-Acetylbenzo-18-crown-6 (4'-acetylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0% (by GC)
Irritant
4'-Acetylbenzo-18-crown-6 (4'-acetylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0% (by GC)
Catalog # TCIAA1604-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4'-Acetylbenzo-18-crown-6 (4'-acetylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97.0% (by GC)
Catalog # TCIAA1604-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Melting point:
    77.0 to 81.0 deg-C

Specification Test Results

About this item

 :
 :GHS07
CAS Number:41855-35-0
Formula:C₁₈H₂₆O₇
MDL Number:MFCD00188456