
4'-Bromobenzo-18-crown-6 (4'-bromobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:75460-28-5
:
Formula:C₁₆H₂₃BrO₆
MDL Number:MFCD00143200
:391.0000 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Melting point:78.0 to 82.0 deg-C
Specification Test Results
Frequently Bought Together