Order Entry
Italy
ContactUsLinkComponent
N-Isopropyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Flammable
Corrosive
N-Isopropyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0420-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
N-Isopropyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Catalog # TCIAI0420-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:162°C
Flash Pt:54°C
EINECS:222-127-2
UN:2734
ADR:8,II
 :GHS02
 :GHS05
CAS Number:3360-16-5
Formula:C₆H₁₆N₂
MDL Number:MFCD00008208