Order Entry
Italy
ContactUsLinkComponent
 
(2-Chloro-3-pyridinyl)methanol ≥98.0% (by GC)
 :  TCI
 
undefined
(2-Chloro-3-pyridinyl)methanol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Gray to Brown powder to lump
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    61.0 to 65.0 deg-C

 

Frequently Bought Together