
4,4'-Dimethoxytrityl chloride ≥97.0% (by HPLC, titration analysis) hydroxyl protecting agent
Supplier: TCI
Certificates
About this item
CAS Number:40615-36-9
:
Formula:C₂₁H₁₉ClO₂
MDL Number:MFCD00008409
:338.8 g/mol
EINECS:255-002-6
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:123.0 to 127.0 deg-C
Specification Test Results
Frequently Bought Together