Order Entry
Italy
ContactUsLinkComponent
(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Irritant
(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Codice catalogo TCIAE0421-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(4-Ethoxybenzyl)triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Codice catalogo TCIAE0421-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    223.0 to 227.0 deg-C

Specification Test Results

About this item

 :
 :477.370 g/mol
 :GHS07
CAS Number:82105-88-2
Formula:C₂₇H₂₆BrOP
MDL Number:MFCD00011834