

2-(Trimethylsiloxy)-1,3-butadiene ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:38053-91-7
:
Formula:C₇H₁₄OSi
MDL Number:MFCD00009852
:142.27 g/mol
Boiling Pt:55°C
Flash Pt:12°C
Storage Temperature:Refrigerator
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together