Order Entry
Italy
ContactUsLinkComponent
 
 
3,5-Di-tert-butyl-2-hydroxybenzaldehyde ≥98.0% (by GC)
 :  TCI
 
undefined
 
3,5-Di-tert-butyl-2-hydroxybenzaldehyde ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Light yellow to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    58.0 to 63.0 deg-C

 

Frequently Bought Together