

Glutaryl dichloride ≥95.0% (by GC, titration analysis)
TCIAG0202-25ML
: TCI
:
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
- Purity (Argentmetric Titration):min. 95.0 %
:
:169.0059 g/mol
:218°C
:106°C
:220-711-1
:2922
:8,II
:GHS05
:GHS06
:2873-74-7
:C₅H₆Cl₂O₂
:MFCD00000755