
1,8-Bis(dimethylamino)naphthalene ≥98.0% (by GC, titration analysis) for dehydrohalogenation
Supplier: TCI
Certificates
About this item
CAS Number:20734-58-1
:
Formula:C₁₄H₁₈N₂
MDL Number:MFCD00003920
:214.31 g/mol
Merck Index:14,03388
Boiling Pt:145°C
EINECS:244-001-6
ADR:8,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:47.0 to 51.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together