Order Entry
Italy
ContactUsLinkComponent
Ethylmagnesium bromide 39% (w/w) ∼3 mol/L in ethyl ether
Flammable
Corrosive
Irritant
Health
Ethylmagnesium bromide 39% (w/w) ∼3 mol/L in ethyl ether
Catalog # TCIAE0134-250G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Ethylmagnesium bromide 39% (w/w) ∼3 mol/L in ethyl ether
Catalog # TCIAE0134-250G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Concentration (sec Butanol method):
    39.0 to 44.0 %

Specification Test Results

About this item

 :
 :133.27 g/mol
EINECS:213-127-3
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:925-90-6
Formula:C₂H₅BrMg
MDL Number:MFCD00000043