Order Entry
Italy
ContactUsLinkComponent
1,7-Diaza-12-crown-4 (1,7-dioxa-4,10-diazacyclododecane) ≥96.0% (by GC, titration analysis)
Irritant
1,7-Diaza-12-crown-4 (1,7-dioxa-4,10-diazacyclododecane) ≥96.0% (by GC, titration analysis)
Catalog # TCIAD2743-100MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,7-Diaza-12-crown-4 (1,7-dioxa-4,10-diazacyclododecane) ≥96.0% (by GC, titration analysis)
Catalog # TCIAD2743-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 96.0 %
  • Melting point:
    85.0 to 90.0 deg-C

Specification Test Results

About this item

 :
 :174.24 g/mol
EINECS:206-034-4
 :GHS07
CAS Number:294-92-8
Formula:C₈H₁₈N₂O₂
MDL Number:MFCD00142571