Order Entry
Italy
ContactUsLinkComponent
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane) ≥98.0% (by titrimetric analysis)
Irritant
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane) ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane) ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    53.0 to 59.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together