Order Entry
Italy
ContactUsLinkComponent
3,3-Diethoxypropiononitrile ≥95.0% (by GC)
Irritant
3,3-Diethoxypropiononitrile ≥95.0% (by GC)
Catalog # TCIAC1178-10ML
Supplier:  TCI
CAS Number:  
undefined
Irritant
3,3-Diethoxypropiononitrile ≥95.0% (by GC)
Catalog # TCIAC1178-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :143.19 g/mol
Boiling Pt:99°C
Flash Pt:84°C
EINECS:217-988-6
 :GHS07
CAS Number:2032-34-0
Formula:C₇H₁₃NO₂
MDL Number:MFCD00040558