Order Entry
Italy
ContactUsLinkComponent
Tetrahydrofuran, anhydrous ≥99.5% (by GC) stabilised
Flammable
Irritant
Health
Tetrahydrofuran, anhydrous ≥99.5% (by GC) stabilised
Catalog # TCIAT2394-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
Tetrahydrofuran, anhydrous ≥99.5% (by GC) stabilised
Catalog # TCIAT2394-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Water:
    max. 50 ppm

Specification Test Results

About this item

 :
 :72.11 g/mol
Merck Index:14,09211
Boiling Pt:65°C
Melting Pt:–108°C
Density:0,888 g/cm³ (20 °C)
Flash Pt:–15°C
Storage Temperature:Ambient
EINECS:203-726-8
UN:2056
ADR:3,II
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
REACH:05-2114615188-48
 :GHS02
 :GHS07
 :GHS08
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356