Order Entry
Italy
ContactUsLinkComponent
Sodium cyanotrihydroborate ≥95.0% (by titrimetric analysis) reducing agent
Flammable
Corrosive
Toxic
Environment
Sodium cyanotrihydroborate ≥95.0% (by titrimetric analysis) reducing agent
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Toxic
Environment
Sodium cyanotrihydroborate ≥95.0% (by titrimetric analysis) reducing agent
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 95.0 %

Specification Test Results

Frequently Bought Together