



Methylcyclohexane ≥99.0% (by GC) for spectrophotometry
Catalog # TCIAM0244-100ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
- Absorbance (210nm):max. 1.000
- Absorbance (220nm):max. 0.500
- Absorbance (230nm):max. 0.200
Specification Test Results
About this item
:
:98.19 g/mol
Merck Index:14,06047
Boiling Pt:100°C
Melting Pt:-127°C
Melting Pt:–127°C
Density:0,7693 g/cm³ (20 °C)
Flash Pt:-4°C
Flash Pt:–4°C
Storage Temperature:Ambient
EINECS:203-624-3
UN:2296
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:108-87-2
Formula:C₆H₁₁CH₃
MDL Number:MFCD00001497