
2-Bromo-6-formylpyridine ≥98.0% (by GC)
: TCI
:34160-40-2
:
:C₆H₄BrNO
:MFCD02683546
:186.01 g/mol
:GHS07
:Refrigerator
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:79.0 to 83.0 deg-C
Frequently Bought Together