
Diethyl hexafluoroglutarate ≥98.0% (by GC)
TCIAH0744-1G
: TCI
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:Light orange to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
:
:296.17 g/mol
:84°C
:87°C
:207-035-2
:GHS07
:424-40-8
:C₉H₁₀F₆O₄
:MFCD00015156