D-Cycloserine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:68-41-7
:
Formula:C₃H₆N₂O₂
MDL Number:MFCD00005353
:102.09 g/mol
Merck Index:14,02751
EINECS:200-688-4
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:+110 to +130 deg (C=1, H2O)
Specification Test Results
Frequently Bought Together