


1,4-Hexadiene cis- and trans-mixture ≥99.0% (by GC)
Catalog # TCIAH0333-5ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
Specification Test Results
About this item
:
:82.15 g/mol
Boiling Pt:65°C
Flash Pt:-21°C
Flash Pt:–21°C
Storage Temperature:Refrigerator
EINECS:209-756-8
UN:2458
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:592-45-0
Formula:C₆H₁₀
MDL Number:MFCD00009298