Order Entry
Italy
ContactUsLinkComponent
Methanol ≥99.8% (by GC) for spectrophotometry
Flammable
Toxic
Health
Methanol ≥99.8% (by GC) for spectrophotometry
Catalog # TCIAM0097-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol ≥99.8% (by GC) for spectrophotometry
Catalog # TCIAM0097-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.8 %
  • Absorbance (210nm):
    max. 0.700
  • Absorbance (220nm):
    max. 0.400
  • Absorbance (230nm):
    max. 0.200

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:14,05957
Boiling Pt:64°C
Melting Pt:–98 °C
Density:0,7918 g/cm³ (20 °C)
Flash Pt:12°C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595