

2-Iodobutane ≥98.0% (by GC) stabilised
: TCI
:513-48-4
:
:C₄H₉I
:MFCD00001072
:184.02 g/mol
:14,01574
:120°C
:–104 °C
:1,6263 g/cm³ (0 °C)
:24°C
:2390
:3,II
:Stabilised with copper chip
:208-163-1
:GHS02
:GHS07
:Ambient
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
Frequently Bought Together