
(4-Carboxybutyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:17814-85-6
:
Formula:C₂₃H₂₄BrO₂P
MDL Number:MFCD00011906
:443.32 g/mol
Melting Pt:205…208 °C
EINECS:241-782-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:205.0 to 209.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together