
1-Bromoperfluoroheptane ≥97.0% (by GC)
Catalog # TCIAP1753-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
- Specific gravity (20/20):1.9170 to 1.9210
- Refractive index n20/D:1.3000 to 1.3060
Specification Test Results
About this item
:
:448.96 g/mol
Boiling Pt:118°C
EINECS:206-799-4
:GHS07
CAS Number:375-88-2
Formula:BrF₂C(CF₂)₅CF₃
MDL Number:MFCD00013570