4-tert-Butylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)
: TCI
:98-52-2
:
:C₁₀H₂₀O
:MFCD00001473
:156.27 g/mol
:214 °C (1013 hPa)
:57 °C
:0,87 g/cm³ (80 °C)
:Refrigerator
:202-676-4
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 % (total of isomers)
- Melting point:64.0 to 71.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together