Tetradecyltrimethylammonium chloride ≥98.0% (by titrimetric analysis)
: TCI
: TTAC, Tetradonium chloride, N,N,N-Trimethyltetradecan-1-aminium chloride, Trimethyl-tetradecylammonium chloride, Myristyltrimethylammonium chloride, n-Tetradecyltrimethylammonium chloride, (1-Tetradecyl)trimethylammonium chloride
:4574-04-3
:
:C₁₇H₃₈ClN
:MFCD00059968
:291.95 g/mol
:224-958-6
:GHS07
- Environmentally Preferable:
- Appearance:White to Almost white powder to crystaline
- Purity (Nonaqueous Titration):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together