PIPES monosodium salt ≥99.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAP0875-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
Specification Test Results
About this item
:
:324.35 g/mol
Merck Index:14,07479
Storage Temperature:Ambient
EINECS:233-005-3
CAS Number:10010-67-0
Formula:C₈H₁₇N₂NaO₆S₂
MDL Number:MFCD00065472