
ω,4-Dibromoacetophenone ≥99.0% (by titrimetric analysis) for HPLC labeling
TCIAA5501-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 99.0 %
- Melting point:109.0 to 112.0 deg-C
- Solubility in Toluene:almost transparency
- Effect test of HPLC derivatization:Effective as labeling agent for Hexanoic Acid.
:
:277.94 g/mol
:14,01427
:310 °C (1013 hPa)
:107…108 °C
:Refrigerator
:202-783-6
:3261
:8,III
:GHS05
:99-73-0
:C₈H₆Br₂O
:MFCD00000200