Order Entry
Italy
ContactUsLinkComponent
3-Pyrroline ≥70% (by GC) (contains Pyrrolidine)
Flammable
Corrosive
3-Pyrroline ≥70% (by GC) (contains Pyrrolidine)
Catalog # TCIAP0577-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
3-Pyrroline ≥70% (by GC) (contains Pyrrolidine)
Catalog # TCIAP0577-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 70.0 %

Specification Test Results

About this item

 :
 :69.11 g/mol
Merck Index:14,08017
Boiling Pt:91°C
Melting Pt:–71 °C
Density:0,9097 g/cm³ (20 °C)
Flash Pt:-18°C
Flash Pt:–18°C
EINECS:203-723-1
UN:2924
ADR:3,II
 :GHS02
 :GHS05
CAS Number:109-96-6
Formula:C₄H₇N
MDL Number:MFCD00005213