


N,N-Dimethylformamide dimethyl acetal for esterification (0.5 mL x 10)
Catalog # TCIAD1332-1SET
Supplier: TCI
CAS Number:
Specifications
- Pk:1 SET
- Seq:0125
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- High boiling impurities:to pass test
Specification Test Results
About this item
:
:119.16 g/mol
Boiling Pt:102 °C (1013 hPa)
Density:0,893 g/cm³ (20 °C)
Flash Pt:5°C
EINECS:225-063-3
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:4637-24-5
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482