Order Entry
Italy
ContactUsLinkComponent
Diethyl oxalate ≥99.0% (by GC) for spectrophotometry
 
Diethyl oxalate ≥99.0% (by GC) for spectrophotometry
  TCIAO0120-250ML
 :  TCI
 :  
undefined
 
Diethyl oxalate ≥99.0% (by GC) for spectrophotometry
  TCIAO0120-250ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

 

 

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Absorbance (320nm):
    max. 0.500
  • Absorbance (330nm):
    max. 0.150
  • Absorbance (340nm):
    max. 0.070

 

 

 :
 :146.14 g/mol
 :14,03125
 :185°C
 :-41°C
 :–41°C
 :1,0785 g/cm³ (20 °C)
 :76°C
 :Ambient
 :202-464-1
 :2525
 :6.1,III
 :GHS07
 :95-92-1
 :C₆H₁₀O₄
 :MFCD00009119