Order Entry
Italy
ContactUsLinkComponent
 
D-(+)-Galactose, anhydrous ≥98.0% (by HPLC)
 :  TCI
 
undefined
D-(+)-Galactose, anhydrous ≥98.0% (by HPLC)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (HPLC) (CAD):
    min. 98.0 area%
  • Specific rotation [a]20/D:
    +79.0 to +81.0 deg (C=10, H2O)
  • Solubility in Water:
    very faint turbidity

 

Frequently Bought Together