
2,4-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:68716-47-2
:
Formula:C₆H₅BCl₂O₂
MDL Number:MFCD00013930
:190.82 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specification Test Results
Related Information
Frequently Bought Together