

4-Chloro-1,1-dimethoxybutane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:29882-07-3
:MFCD01632883
Formula:C₆H₁₃ClO₂
:152.62 g/mol
Boiling Pt:120°C
Flash Pt:43°C
EINECS:249-924-8
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together