Order Entry
Italy
ContactUsLinkComponent
(1-Tetradecyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Irritant
(1-Tetradecyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1506-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
(1-Tetradecyl)triphenylphosphonium bromide ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAT1506-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Water:
    almost transparency

Specification Test Results

About this item

 :
 :539.58 g/mol
EINECS:247-267-1
 :GHS07
CAS Number:25791-20-2
Formula:H₃C(CH₂)₁₃P(C₆H₅)₃Br
MDL Number:MFCD00051857