
3-Formylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:87199-16-4
:
Formula:C₇H₇BO₃
MDL Number:MFCD00161356
:149.94 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):97.0 to 114.0 %
Specification Test Results
Related Information
Frequently Bought Together