Order Entry
Italy
ContactUsLinkComponent
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture ≥97.0% (by GC)
Flammable
Corrosive
Toxic
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture ≥97.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Toxic
Health
Environment
Decahydronaphthalene (mixture of cis and trans isomers) cis- and trans-mixture ≥97.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 % (total of isomer)

Specification Test Results

Frequently Bought Together