

(Diethylamino)sulphur trifluoride ≥90.0% (by titrimetric analysis) fluorinating reagent
Supplier: TCI
Certificates
About this item
CAS Number:38078-09-0
:
Formula:C₄H₁₀F₃NS
MDL Number:MFCD00000363
:161.19 g/mol
Merck Index:14,03113
Boiling Pt:32°C
Density:1,316 g/cm³ (20 °C)
Flash Pt:23°C
Storage Temperature:Refrigerator
EINECS:253-771-2
UN:2920
ADR:8,I
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (Neutralization titration):min. 90.0 %
Specification Test Results
Frequently Bought Together