Order Entry
Italy
ContactUsLinkComponent
1-(2-Chloroethoxy)propane ≥97.0% (by GC)
Flammable
Irritant
1-(2-Chloroethoxy)propane ≥97.0% (by GC)
Catalog # TCIAC1174-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
1-(2-Chloroethoxy)propane ≥97.0% (by GC)
Catalog # TCIAC1174-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :122.59 g/mol
Boiling Pt:129°C
Melting Pt:–60 °C
Density:0,98721 g/cm³ (0 °C)
Flash Pt:29°C
EINECS:255-685-0
UN:3271
ADR:3,III
 :GHS02
 :GHS07
CAS Number:42149-74-6
Formula:ClCH₂CH₂OCH₂CH₂CH₃
MDL Number:MFCD00059052