Reagent kit for the detection of glucans, GLUCATELL®
Certificates
About this item
GLUCATELL® is a β-D-glucan-specific chromogenic reagent. The products listed below are offered for research use only. These products are not licensed for use in FDA regulated endotoxin detection studies or for diagnostic purposes. Each kit contains one respectively two vials each of GLUCATELL®-Reagent (55 tests/vial), reconstitution buffer, (1—>3)-β-D-glucan-standard, reagent water and one or two microplates. Kits for the endpoint assay also contain reagents for diazo-coupling (540 to 550 nm).
Frequently Bought Together