Order Entry
Italy
ContactUsLinkComponent
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥93.0% (by titrimetric analysis)
Irritant
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥93.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Chloro(1,5-cyclooctadiene)iridium(I) dimer ≥93.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Amber to Dark red powder to crystal
  • Purity (Argentmetric Titration):
    min. 93.0 %

Specification Test Results

Related Information

Frequently Bought Together