Bis(1,5-cyclooctadiene)iridium(I) tetrafluoroborate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:35138-23-9
:
Formula:C₁₆H₂₄BF₄Ir
MDL Number:MFCD00079664
:495.383 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together