Order Entry
Italy
ContactUsLinkComponent
Dansyl chloride 10% in acetone
Flammable
Corrosive
Irritant
Health
Dansyl chloride 10% in acetone
Catalog # TCIAD0005-10ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Dansyl chloride 10% in acetone
Catalog # TCIAD0005-10ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown clear liquid
  • Concentration (Nonaqueous Titration):
    10.0 to 11.5 w/w%

Specification Test Results

About this item

 :
 :269.75 g/mol
Merck Index:14,02814
Storage Temperature:Refrigerator
EINECS:210-092-6
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:605-65-2
Formula:C₁₂H₁₂ClNO₂S
MDL Number:MFCD00003985