
1-Cyclohexene-1-carboxylic acid 96%
Catalog # ACRO291750025
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 g
- Seq:3122
Specifications
Specification Test Results
- Appearance:White to light yellow Crystalline mass or liquid
- Infrared spectrum:Conforms
- HPLC:≥95 %
Specification Test Results
About this item
:
:126.16 g/mol
Boiling Pt:137 °C (15 mmHg)
Melting Pt:35…39 °C
ADR:8,III
:GHS07
Storage Temperature:Ambient
CAS Number:636-82-8
Formula:C₇H₁₀O₂
MDL Number:MFCD00001545