
3-Amino-4-(methylthio)benzotrifluoride ≥98.0% (by GC, titration analysis)
Catalog # TCIAA1666-1G
Supplier: TCI
CAS Number:
Specifications
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
Boiling Pt:112°C
ADR:6.1,III
:207.22 g/mol
:GHS07
CAS Number:207974-07-0
Formula:C₈H₈F₃NS
MDL Number:MFCD00052715