N-Acetyl-L-leucine ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1188-21-2
:
Formula:C₈H₁₅NO₃
MDL Number:MFCD00065131
:173.2 g/mol
EINECS:214-706-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Melting point:179.0 to 186.0 deg-C
- Specific rotation [a]20/D:-20.0 to -24.0 deg (C=5, EtOH)
- Solubility in EtOH:almost transparency
Specification Test Results
Frequently Bought Together