Order Entry
Italy
ContactUsLinkComponent
 
N-Acetyl-D-leucine ≥99.0% (by titrimetric analysis)
  TCIAA0713-1G
 :  TCI
 :  
undefined
N-Acetyl-D-leucine ≥99.0% (by titrimetric analysis)
  TCIAA0713-1G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 99.0 %
  • Specific rotation [a]20/D:
    +20.0 to +24.0 deg (C=5, EtOH)
  • Solubility in EtOH:
    almost transparency

 

 

 :
 :173.2 g/mol
 :19764-30-8
 :C₈H₁₅NO₃
 :MFCD00066069