
β-Bromostyrene (mixture of cis and trans isomers) cis- and trans-mixture ≥95.0% (by GC)
TCIAB0655-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Yellow clear liquid
- Purity (GC):min. 95.0% (total of isomer)
:
:183.05 g/mol
:120°C
:203-131-3
:GHS07
:Refrigerator
:103-64-0
:C₈H₇Br
:MFCD00000185